Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_172894.2 | NP_766482.2 | 5, 6 | 1099 | |
GenBank mRNA | AK080891.1 | - | 5, 6 | 1140 | |
AK122450.1 | - | 5, 6 | 1099 | ||
BC053076.1 | - | 5, 6 | 1099 | 5698212 | |
BC141039.1 | - | 4, 5 | 646 | 9055651 |