Gene Transcripts

Gene Symbol
Npat
Entrez Gene ID
244879
Gene Name
nuclear protein in the AT region
Gene Aliases
6820401K01, AI427561, BB112559
Chromosome Location
Chr. 9: 53537041 - 53575627 on Build GRCm38
UniGene ID
Mm.116713
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081152.1 NP_001074621.1 17 4186
GenBank mRNA AK042624.1 - 1 859
AK053074.1 - 1 835
AK080294.1 - 17 4194
BC024717.1 - 1 208

View Full Product Details