Gene Transcripts

Gene Symbol
Peak1
Entrez Gene ID
244895
Gene Name
pseudopodium-enriched atypical kinase 1
Gene Aliases
1110049L02Rik, 9530046P14, AA690169, BE132841, C230081A13Rik, SGK269, mKIAA2002
Chromosome Location
Chr. 9: 56201126 - 56418050 on Build GRCm38
UniGene ID
Mm.490331
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172924.3 NP_766512.2 4 1108
GenBank mRNA AK035440.1 - 4 1098
AK048912.1 - 4 1097
AK090133.1 - 4 1124

View Full Product Details