Gene Transcripts

Gene Symbol
Slitrk2
Entrez Gene ID
245450
Gene Name
SLIT and NTRK-like family, member 2
Gene Aliases
A930040J07
Chromosome Location
Chr. X: 66649318 - 66661402 on Build GRCm38
UniGene ID
Mm.336081
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001161431.1 NP_001154903.1 3 2562
NM_198863.2 NP_942563.2 2 2538
GenBank mRNA BC111888.1 - 2 2537
BC112406.1 - 2 2537 40060552

View Full Product Details