Gene Transcripts

Gene Symbol
Hsf3
Entrez Gene ID
245525
Gene Name
heat shock transcription factor 3
Gene Aliases
B230358A15Rik, HSF 3, HSTF 3, mHSF3
Chromosome Location
Chr. X: 96306184 - 96367919 on Build GRCm38
UniGene ID
Mm.166749
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172931.3 NP_766519.1 8 782
GenBank mRNA AB510446.1 - 9 884
AB510447.1 - 1384
AK046229.1 - 812
BC117805.2 - 8 782 40092308

View Full Product Details