Gene Transcripts

Gene Symbol
Onecut3
Entrez Gene ID
246086
Gene Name
one cut domain, family member 3
Gene Aliases
OC-3, Oc3
Chromosome Location
Chr. 10: 80494906 - 80517266 on Build GRCm38
UniGene ID
Mm.250572
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_139226.3 NP_631972.2 1, 2 1294
GenBank mRNA AY080897.1 - 1, 2 1245
BC058700.1 - 1, 2 1294 6850438

View Full Product Details