Gene Transcripts

Gene Symbol
PRKX
Entrez Gene ID
5613
Gene Name
protein kinase, X-linked
Gene Aliases
PKX1
Chromosome Location
Chr. X: 3604343 - 3713634 on Build GRCh38
UniGene ID
Hs.390788
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005044.4 NP_005035.1 6 1223
GenBank mRNA BC041073.1 - 6 1026
X85545.1 - 6 1208

Gene Symbol
PRKXP1
Entrez Gene ID
441733
Gene Name
protein kinase, X-linked, pseudogene 1
Gene Aliases
Chromosome Location
Chr. 15: 100547752 - 100559283 on Build GRCh38
UniGene ID
Hs.12250
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_073405.1 5520

View Full Product Details