Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000313.3 | NP_000304.2 | 12 | 1798 | |
NM_001314077.1 | NP_001301006.1 | 13 | 1885 | ||
GenBank mRNA | AK292994.1 | - | 12 | 1604 | |
AK303895.1 | - | 5 | 725 | ||
BC015801.1 | - | 12 | 1774 | 3909023 | |
KU178283.1 | - | 12 | 1457 | ||
KU178284.1 | - | 13 | 1553 | ||
M14338.1 | - | 12 | 1625 | ||
M15036.1 | - | 12 | 1603 | ||
X12892.1 | - | 12 | 1625 | ||
Y00692.1 | - | 12 | 1580 |