Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_002775.4 | NP_002766.1 | 7 | 1276 | |
GenBank mRNA | AF070555.1 | - | 5 | 550 | |
AK091944.1 | - | 9 | 1051 | ||
AK092476.1 | - | 8 | 1057 | ||
AK290089.1 | - | 7 | 767 | ||
BC011352.1 | - | 8 | 1043 | ||
BC031082.1 | - | 7 | 1184 | ||
D87258.1 | - | 7 | 1196 | ||
Y07921.1 | - | 7 | 1186 |