Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_170597.4 | NP_733485.1 | 4 | 1048 | |
GenBank mRNA | AB046110.1 | - | 4 | 1366 | |
AK034501.1 | - | 4 | 811 | ||
AK043871.1 | - | 4 | 813 | ||
AK044192.1 | - | 4 | 1048 | ||
AK046245.1 | - | 4 | 835 | ||
AK134713.1 | - | 4 | 819 | ||
AK134861.1 | - | 4 | 1109 | ||
BC034115.1 | - | 4 | 736 | ||
BC116790.1 | - | 4 | 941 | 40126109 | |
BC116792.1 | - | 4 | 941 | 40126111 |