Gene Transcripts

Gene Symbol
Klhl18
Entrez Gene ID
270201
Gene Name
kelch-like 18
Gene Aliases
A930041K15, AW545966
Chromosome Location
Chr. 9: 110425926 - 110476700 on Build GRCm38
UniGene ID
Mm.41744
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177771.5 NP_808439.2 3 384
GenBank mRNA AK044774.1 - 3 495
BC065167.1 - 3 280
BY195998.1 - 3 305
BY248410.1 - 3 304

View Full Product Details