Gene Transcripts

Gene Symbol
AEBP1
Entrez Gene ID
165
Gene Name
AE binding protein 1
Gene Aliases
ACLP
Chromosome Location
Chr. 7: 44104320 - 44114565 on Build GRCh38
UniGene ID
Hs.439463
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001129.4 NP_001120.3 19 2895
XM_011515162.1 XP_011513464.1 18 2858
GenBank mRNA AB209744.1 - 19 2849
AF053944.1 - 19 2729
AK090931.1 - 6 987
AK127541.1 - 6 1383
AK296001.1 - 5 889
BC038588.1 - 19 2895 5764769
D86479.1 - 13 1651

View Full Product Details