Gene Transcripts

Gene Symbol
Fam26d
Entrez Gene ID
270711
Gene Name
family with sequence similarity 26, member D
Gene Aliases
4732454E20Rik, AI586164, Gm643
Chromosome Location
Chr. 10: 34038784 - 34044313 on Build GRCm38
UniGene ID
Mm.303077
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081165.1 NP_001074634.1 1 146
GenBank mRNA AK028763.1 - 1 140

View Full Product Details