Gene Transcripts

Gene Symbol
Wnk3
Entrez Gene ID
279561
Gene Name
WNK lysine deficient protein kinase 3
Gene Aliases
Prkwnk3, Wnk3-ps
Chromosome Location
Chr. X: 151168777 - 151320194 on Build GRCm38
UniGene ID
Mm.441474
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271678.1 NP_001258607.1 22 5149
NM_001271679.1 NP_001258608.1 22 5008
GenBank mRNA BC043119.1 - 5 1202
BC060731.1 - 5 1202
JQ247189.1 - 22 5149
JQ247190.1 - 22 5008

View Full Product Details