Gene Transcripts

Gene Symbol
Hist1h2be
Entrez Gene ID
319179
Gene Name
histone cluster 1, H2be
Gene Aliases
Chromosome Location
Chr. 13: 23583670 - 23621124 on Build GRCm38
UniGene ID
Mm.440391
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001177653.1 NP_001171124.1 2 1221
NM_178194.4 NP_835501.1 2 1247
GenBank mRNA AK030546.1 - 2 1249
AK049948.1 - 2 1222

View Full Product Details