Gene Transcripts

Gene Symbol
Efcab5
Entrez Gene ID
319634
Gene Name
EF-hand calcium binding domain 5
Gene Aliases
4930563A03Rik
Chromosome Location
Chr. 11: 77089904 - 77199943 on Build GRCm38
UniGene ID
Mm.108786
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_176965.3 NP_795939.2 12 3139
GenBank mRNA AK029790.1 - 10 2726
AK035698.1 - 8 1973
BC127154.1 - 10 2681

View Full Product Details