Gene Transcripts

Gene Symbol
Bbs9
Entrez Gene ID
319845
Gene Name
Bardet-Biedl syndrome 9 (human)
Gene Aliases
E130103I17Rik
Chromosome Location
Chr. 9: 22475542 - 22888283 on Build GRCm38
UniGene ID
Mm.176725
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178415.1 NP_848502.1 16 1961
NM_181316.4 NP_851833.2 15 1905
GenBank mRNA AK028200.1 - 15 1995
AK086114.1 - 15 1877
AK144730.1 - 16 1932
AK166297.1 - 15 1886
BC030419.1 - 13 1269
BC046426.1 - 15 1917 3601505
BC054441.1 - 4 164

View Full Product Details