Gene Transcripts

Gene Symbol
Cep95
Entrez Gene ID
320162
Gene Name
centrosomal protein 95
Gene Aliases
4732496G21Rik, AI118346, AI448335, Ccdc45, D330027A14Rik, F630025I20Rik
Chromosome Location
Chr. 11: 106789252 - 106818861 on Build GRCm38
UniGene ID
Mm.30524
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177088.3 NP_796062.2 6, 7 690
GenBank mRNA AK052705.1 - 6, 7 618
AK076400.1 - 6, 7 668
BC090675.1 - 6, 7 648

View Full Product Details