Gene Transcripts

Gene Symbol
Fnip2
Entrez Gene ID
329679
Gene Name
folliculin interacting protein 2
Gene Aliases
D630023B12Rik, mKIAA1450
Chromosome Location
Chr. 3: 79455971 - 79567879 on Build GRCm38
UniGene ID
Mm.244005
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001162999.2 NP_001156471.1 3 412

View Full Product Details