Gene Transcripts

Gene Symbol
Acad12
Entrez Gene ID
338350
Gene Name
acyl-Coenzyme A dehydrogenase family, member 12
Gene Aliases
9330129D05Rik
Chromosome Location
Chr. 5: 121598277 - 121618938 on Build GRCm38
UniGene ID
Mm.482918
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178799.3 NP_848914.2 6 1098
GenBank mRNA AK033963.1 - 6 1017
AK080793.1 - 6 1098

View Full Product Details