Gene Transcripts

Gene Symbol
Wee2
Entrez Gene ID
381759
Gene Name
WEE1 homolog 2 (S. pombe)
Gene Aliases
Gm1065, Wee1B
Chromosome Location
Chr. 6: 40426699 - 40466815 on Build GRCm38
UniGene ID
Mm.20593
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_201370.2 NP_958758.2 6 1179
GenBank mRNA AK139573.1 - 6 1138
AK143336.1 - 6 1103
AK163344.1 - 6 1181
BC052883.1 - 6 1086 30053490
BC080784.1 - 6 1104 30481521
DQ011691.1 - 6 1000

View Full Product Details