Gene Transcripts

Gene Symbol
Ripply2
Entrez Gene ID
382089
Gene Name
ripply2 homolog (zebrafish)
Gene Aliases
C030002E08Rik, Gm1122
Chromosome Location
Chr. 9: 87015128 - 87020441 on Build GRCm38
UniGene ID
Mm.92244
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037907.1 NP_001032996.1 4 265
GenBank mRNA AB212223.1 - 4 265
AK081203.1 - 3 407

View Full Product Details