Gene Transcripts

Gene Symbol
Tas2r125
Entrez Gene ID
387352
Gene Name
taste receptor, type 2, member 125
Gene Aliases
T2R26, Tas2r25, mGR25, mt2r59
Chromosome Location
Chr. 6: 132909651 - 132910586 on Build GRCm38
UniGene ID
Mm.377913
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207027.1 NP_996910.1 1 644
GenBank mRNA BC107190.1 - 1 644
BC107191.1 - 1 644 40050841

View Full Product Details