Gene Transcripts

Gene Symbol
RBMS1
Entrez Gene ID
5937
Gene Name
RNA binding motif single stranded interacting protein 1
Gene Aliases
C2orf12, HCC-4, MSSP, MSSP-1, MSSP-2, MSSP-3, SCR2, YC1
Chromosome Location
Chr. 2: 160272151 - 160493856 on Build GRCh38
UniGene ID
Hs.470412
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002897.4 NP_002888.1 8 1225
NM_016836.3 NP_058520.1 8 1234
XM_005246737.1 XP_005246794.1 8 945
XM_005246738.2 XP_005246795.1 8 1280
XM_005246739.2 XP_005246796.1 8 1150
XM_005246740.1 XP_005246797.1 8 936
XM_005246741.3 XP_005246798.1 8 1147
XM_017004626.1 XP_016860115.1 8 1271
GenBank mRNA AK225887.1 - 8 845
AK297812.1 - 8 936
AK309949.1 - 8 909
BC012992.2 - 8 920 3530897
BC012993.2 - 8 920 3530898
BC018951.2 - 8 1221 4137722
BC051889.1 - 8 807
BX648246.1 - 2 4982
D28482.1 - 8 791
KU178305.1 - 8 781
KU178306.1 - 8 787
L11289.1 - 8 858
X64652.1 - 7 743
X77494.1 - 9 922

View Full Product Details