Gene Transcripts

Gene Symbol
Ccdc87
Entrez Gene ID
399599
Gene Name
coiled-coil domain containing 87
Gene Aliases
4931419P11Rik
Chromosome Location
Chr. 19: 4839366 - 4842528 on Build GRCm38
UniGene ID
Mm.444704
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207268.3 NP_997151.2 1 1755
GenBank mRNA AK029877.1 - 1 1707
BC145660.1 - 1 1660 40130963

View Full Product Details