Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_020485.4 | NP_065231.3 | 10 | 1504 | |
NM_138616.3 | NP_619522.3 | 7 | 1051 | ||
NM_138617.3 | NP_619523.3 | 7 | 1109 | ||
NM_138618.3 | NP_619524.3 | 9 | 1370 | ||
GenBank mRNA | JX420291.1 | - | 10 | 1418 | |
JX420292.1 | - | 10 | 1418 | ||
JX420293.1 | - | 10 | 1418 | ||
JX420294.1 | - | 10 | 1418 | ||
JX420295.1 | - | 10 | 1418 | ||
JX420296.1 | - | 10 | 1418 | ||
JX420297.1 | - | 10 | 1418 | ||
JX420298.1 | - | 10 | 1418 | ||
JX420299.1 | - | 10 | 1418 | ||
JX420300.1 | - | 10 | 1418 | ||
JX420301.1 | - | 10 | 1418 | ||
JX420302.1 | - | 10 | 1418 | ||
X63095.1 | - | 7 | 1031 |