Gene Transcripts

Gene Symbol
Zbtb9
Entrez Gene ID
474156
Gene Name
zinc finger and BTB domain containing 9
Gene Aliases
3930402F13Rik
Chromosome Location
Chr. 17: 26973179 - 26976203 on Build GRCm38
UniGene ID
Mm.328890
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005916.2 NP_001005916.1 2 382
GenBank mRNA AK014458.1 - 1 214
AK030262.1 - 2 271
AK154850.1 - 2 364
BC049933.1 - 2 382 5387663

View Full Product Details