Gene Transcripts
- Gene Symbol
- RIT1
- Entrez Gene ID
- 6016
- Gene Name
- Ras like without CAAX 1
- Gene Aliases
- NS8, RIBB, RIT, ROC1
- Chromosome Location
- Chr. 1: 155897808 - 155911402 on Build GRCh38
- UniGene ID
- Hs.491234
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001256820.1 |
NP_001243749.1
|
2, 3 |
217 |
|
NM_001256821.1 |
NP_001243750.1
|
3, 4 |
244 |
|
NM_006912.5 |
NP_008843.1
|
3, 4 |
366 |
|
GenBank mRNA |
AF084462.1 |
- |
3, 4 |
277 |
|
AF493923.1 |
- |
2, 3 |
163 |
|
AK298768.1 |
- |
2, 3 |
217 |
|
AK314239.1 |
- |
3, 4 |
342 |
|
BC104186.2 |
- |
3, 4 |
237 |
40030939 |
BC104187.2 |
- |
3, 4 |
237 |
40030940 |
BI916790.1 |
- |
3, 4 |
227 |
|
BM546985.1 |
- |
3, 4 |
360 |
|
BQ918446.1 |
- |
3, 4 |
244 |
|
CR407639.1 |
- |
2, 3 |
163 |
|
U71203.1 |
- |
3, 4 |
298 |
|
U78165.1 |
- |
3, 4 |
298 |
|
Y07566.1 |
- |
3, 4 |
308 |
|
View Full Product Details