Gene Transcripts

Gene Symbol
RNASE1
Entrez Gene ID
6035
Gene Name
ribonuclease A family member 1, pancreatic
Gene Aliases
RAC1, RIB1, RNS1
Chromosome Location
Chr. 14: 20801356 - 20802877 on Build GRCh38
UniGene ID
Hs.78224
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002933.4 NP_002924.1 2 225
NM_198232.2 NP_937875.1 2 295
NM_198234.2 NP_937877.1 3 296
NM_198235.2 NP_937878.1 3 308
GenBank mRNA AK098375.1 - 2 275
AK129871.1 - 2 216
AK312100.1 - 2 196
AV710596.1 - 3 266
BC005324.1 - 2 225 3950230
BC022882.1 - 2 199 4294153
BF106121.1 - 173
BG539567.1 - 2 196
BI599530.1 - 2 197
BI753463.1 - 2 295
BI760598.1 - 2 183
BM007311.1 - 2 168
BM919747.1 - 2 180
CB993343.1 - 3 305
CD703706.1 - 3 296
CR450318.1 - 1 119

View Full Product Details