Gene Transcripts

Gene Symbol
Cyp26c1
Entrez Gene ID
546726
Gene Name
cytochrome P450, family 26, subfamily c, polypeptide 1
Gene Aliases
EG546726
Chromosome Location
Chr. 19: 37685680 - 37693307 on Build GRCm38
UniGene ID
Mm.457750
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105201.1 NP_001098671.1 2 365
GenBank mRNA BC151098.1 - 2 464
BC151106.1 - 2 464 9088019

View Full Product Details