Gene Transcripts

Gene Symbol
Fam71a
Entrez Gene ID
619288
Gene Name
family with sequence similarity 71, member A
Gene Aliases
4933417M04Rik, GARI-L4
Chromosome Location
Chr. 1: 191162584 - 191164817 on Build GRCm38
UniGene ID
Mm.61148
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109759.1 NP_001103229.1 1 1295
GenBank mRNA AB473728.1 - 1 922
AK133412.1 - 1 1297

View Full Product Details