Gene Transcripts
- Gene Symbol
- Rps13
- Entrez Gene ID
- 68052
- Gene Name
- ribosomal protein S13
- Gene Aliases
- 2700063M04Rik
- Chromosome Location
- Chr. 7: 116331507 - 116334190 on Build GRCm38
- UniGene ID
- Mm.14798
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AV498110.1 |
- |
4, 5 |
405 |
|
BC090397.1 |
- |
5, 6 |
425 |
5150054 |
View Full Product Details