Gene Transcripts
- Gene Symbol
- RPL37
- Entrez Gene ID
- 6167
- Gene Name
- ribosomal protein L37
- Gene Aliases
- L37
- Chromosome Location
- Chr. 5: 40831328 - 40835285 on Build GRCh38
- UniGene ID
- Hs.447582
- Species
- Homo sapiens
- Gene Symbol
- SNORD72
- Entrez Gene ID
- 619564
- Gene Name
- small nucleolar RNA, C/D box 72
- Gene Aliases
- HBII-240
- Chromosome Location
- Chr. 5: 40832656 - 40832735 on Build GRCh38
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK124098.1 |
- |
1 |
2665 |
|
View Full Product Details