Gene Transcripts

Gene Symbol
Dact3
Entrez Gene ID
629378
Gene Name
dapper homolog 3, antagonist of beta-catenin (xenopus)
Gene Aliases
EG666671, naa-3
Chromosome Location
Chr. 7: 16875317 - 16887301 on Build GRCm38
UniGene ID
Mm.491123
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081655.1 NP_001075124.1 4 1858
GenBank mRNA AB073971.1 - 4 1777
DQ832319.1 - 4 1858

View Full Product Details