Gene Transcripts
- Gene Symbol
- RPS19
- Entrez Gene ID
- 6223
- Gene Name
- ribosomal protein S19
- Gene Aliases
- DBA, DBA1, S19
- Chromosome Location
- Chr. 19: 41859918 - 41871416 on Build GRCh38
- UniGene ID
- Hs.438429
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001022.3 |
NP_001013.1
|
4, 5 |
739 |
|
NM_001321483.1 |
NP_001308412.1
|
4, 5 |
416 |
|
NM_001321484.1 |
NP_001308413.1
|
4, 5 |
546 |
|
NM_001321485.1 |
NP_001308414.1
|
4, 5 |
752 |
|
XM_017027113.1 |
XP_016882602.1
|
4, 5 |
734 |
|
GenBank mRNA |
AA165492.1 |
- |
2, 3 |
292 |
|
AK311786.1 |
- |
4, 5 |
402 |
|
BC000023.2 |
- |
4, 5 |
436 |
3504083 |
BC007615.2 |
- |
3, 4 |
367 |
3354280 |
BC017386.1 |
- |
4, 5 |
405 |
|
BC018616.1 |
- |
4, 5 |
739 |
4867036 |
BG398397.1 |
- |
4, 5 |
546 |
|
BU603942.1 |
- |
4, 5 |
418 |
|
CB124389.1 |
- |
4, 5 |
445 |
|
CB125125.1 |
- |
4, 5 |
416 |
|
M81757.1 |
- |
4, 5 |
389 |
|
View Full Product Details