Gene Transcripts

Gene Symbol
Adamts5
Entrez Gene ID
23794
Gene Name
a disintegrin-like and metallopeptidase (reprolysin type) with thrombospondin type 1 motif, 5 (aggrecanase-2)
Gene Aliases
9530092O11Rik, ADAM-TS5, ADAMTS1, ADAMTS11, AI481094, ASMP-2
Chromosome Location
Chr. 16: 85858157 - 85902374 on Build GRCm38
UniGene ID
Mm.112933
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011782.2 NP_035912.2 1 1349
GenBank mRNA AF140673.1 - 1 508
BC138619.1 - 1 533 8861639
BC138620.1 - 1 533 8861640

View Full Product Details