Gene Transcripts
- Gene Symbol
- Lzts1
- Entrez Gene ID
- 211134
- Gene Name
- leucine zipper, putative tumor suppressor 1
- Gene Aliases
- F37, FEZ1
- Chromosome Location
- Chr. 8: 69132669 - 69184220 on Build GRCm38
- UniGene ID
- Mm.409446
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK138907.1 |
- |
1 |
69 |
|
View Full Product Details