Gene Transcripts
- Gene Symbol
- Tcra-V22.1
- Entrez Gene ID
- 24078
- Gene Name
- T cell receptor alpha, variable 22.1
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.350840
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
U47051.1 |
- |
2 |
203 |
|
View Full Product Details