Gene Transcripts

Gene Symbol
Igh-VJ558
Entrez Gene ID
16061
Gene Name
immunoglobulin heavy chain (J558 family)
Gene Aliases
AI893585
Chromosome Location
Chr. : - on Build
UniGene ID
Mm.246497
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AF296432.1 - 1 177

View Full Product Details