Gene Transcripts

Gene Symbol
Fbxw28
Entrez Gene ID
668758
Gene Name
F-box and WD-40 domain protein 28
Gene Aliases
Gm9337
Chromosome Location
Chr. 9: 109322886 - 109339659 on Build GRCm38
UniGene ID
Mm.445163
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001177419.1 NP_001170890.1 7 890
NM_001177420.1 NP_001170891.1 7 890
GenBank mRNA AK143322.1 - 7 886
BC084592.1 - 7 881

View Full Product Details