Gene Transcripts

Gene Symbol
Pydc3
Entrez Gene ID
100033459
Gene Name
pyrin domain containing 3
Gene Aliases
9830148g24rik, E430029J22Rik
Chromosome Location
Chr. 1: 173673628 - 173698392 on Build GRCm38
UniGene ID
Mm.303377
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001162938.1 NP_001156410.1 4 1420
GenBank mRNA AK036671.1 - 4 1190

View Full Product Details