Gene Transcripts

Gene Symbol
Shbg
Entrez Gene ID
24775
Gene Name
sex hormone binding globulin
Gene Aliases
Abpa
Chromosome Location
Chr. 10: 56219861 - 56237410 on Build Rnor_6.0
UniGene ID
Rn.37473
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012650.1 NP_036782.1 1 81
XM_006246586.2 XP_006246648.1 2 830
GenBank mRNA BC097336.1 - 1 69
M15034.1 - 1 81
M31179.1 - 1 33

View Full Product Details