Gene Transcripts

Gene Symbol
Scn2b
Entrez Gene ID
25349
Gene Name
sodium channel, voltage-gated, type II, beta subunit
Gene Aliases
SCNB2
Chromosome Location
Chr. 8: 49419003 - 49431110 on Build Rnor_6.0
UniGene ID
Rn.88636
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012877.1 NP_037009.1 2 384
XM_006242895.2 XP_006242957.1 2 336
GenBank mRNA U37026.1 - 2 384

View Full Product Details