Gene Transcripts
- Gene Symbol
- Amelx
- Entrez Gene ID
- 29160
- Gene Name
- amelogenin, X-linked
- Gene Aliases
- Amel
- Chromosome Location
- Chr. X: 26439197 - 26450495 on Build Rnor_6.0
- UniGene ID
- Rn.11368
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_019154.1 |
NP_062027.1
|
3 |
132 |
|
GenBank mRNA |
U67130.1 |
- |
3 |
132 |
|
View Full Product Details