Gene Transcripts

Gene Symbol
SMARCC2
Entrez Gene ID
6601
Gene Name
SWI/SNF related, matrix associated, actin dependent regulator of chromatin subfamily c member 2
Gene Aliases
BAF170, CRACC2, Rsc8
Chromosome Location
Chr. 12: 56161852 - 56189567 on Build GRCh38
UniGene ID
Hs.236030
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001130420.1 NP_001123892.1 10, 11 1073
NM_003075.3 NP_003066.2 10, 11 1073
NM_139067.2 NP_620706.1 10, 11 1073
XM_005269101.1 XP_005269158.1 10, 11 1010
XM_005269102.1 XP_005269159.1 10, 11 1010
XM_005269103.1 XP_005269160.1 10, 11 1010
XM_005269104.1 XP_005269161.1 10, 11 1010
XM_011538693.2 XP_011536995.1 9, 10 919
XM_017019884.1 XP_016875373.1 10, 11 1010
XM_017019885.1 XP_016875374.1 10, 11 1010
XM_017019886.1 XP_016875375.1 10, 11 1010
XM_017019887.1 XP_016875376.1 9, 10 897
GenBank mRNA AB209006.1 - 10, 11 1073
AB209293.1 - 10, 11 982
AK293896.1 - 10, 11 960
AK315153.1 - 10, 11 973
BC013045.1 - 10, 11 999 4180388
BC026222.2 - 10, 11 992 4395012
BT009924.1 - 10, 11 967
U66616.1 - 10, 11 989

View Full Product Details