Gene Transcripts
- Gene Symbol
- Hes2
- Entrez Gene ID
- 29567
- Gene Name
- hes family bHLH transcription factor 2
- Gene Aliases
- Chromosome Location
- Chr. 5: 169338097 - 169339142 on Build Rnor_6.0
- UniGene ID
- Rn.44655
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_019236.1 |
NP_062109.1
|
3 |
342 |
|
GenBank mRNA |
D14029.1 |
- |
3 |
342 |
|
View Full Product Details