Gene Transcripts

Gene Symbol
Timm17a
Entrez Gene ID
54311
Gene Name
translocase of inner mitochondrial membrane 17 homolog A (yeast)
Gene Aliases
Tim17, Timm17
Chromosome Location
Chr. 13: 52124641 - 52136127 on Build Rnor_6.0
UniGene ID
Rn.22514
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019351.1 NP_062224.1 3 162
GenBank mRNA AB006450.1 - 3 162
BC061982.1 - 3 209 5598788

Gene Symbol
Timm17al1
Entrez Gene ID
500655
Gene Name
translocase of inner mitochondrial membrane 17a-like 1
Gene Aliases
RGD1560665
Chromosome Location
Chr. 6: 83252019 - 83253099 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_576033.5 XP_576033.1 2 181

View Full Product Details