Gene Transcripts

Gene Symbol
Tas2r121
Entrez Gene ID
78983
Gene Name
taste receptor, type 2, member 121
Gene Aliases
T2R7, Tas2r13, Tas2r7
Chromosome Location
Chr. 4: 166868482 - 166869399 on Build Rnor_6.0
UniGene ID
Rn.48786
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023997.1 NP_076487.1 1 473
GenBank mRNA AF227144.1 - 1 473

View Full Product Details