Gene Transcripts
- Gene Symbol
- Htr5b
- Entrez Gene ID
- 79247
- Gene Name
- 5-hydroxytryptamine (serotonin) receptor 5B
- Gene Aliases
- 5Ht5b, REC17
- Chromosome Location
- Chr. 13: 37479758 - 37492680 on Build Rnor_6.0
- UniGene ID
- Rn.202942
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_024395.1 |
NP_077371.1
|
1 |
519 |
|
GenBank mRNA |
L10073.1 |
- |
1 |
522 |
|
View Full Product Details