Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000456.2 | NP_000447.2 | 6 | 1846 | |
NM_001032386.1 | NP_001027558.1 | 5 | 1720 | ||
NM_001032387.1 | NP_001027559.1 | 4 | 1611 | ||
XM_005269112.1 | XP_005269169.1 | 5 | 1702 | ||
XM_017019905.1 | XP_016875394.1 | 7 | 2392 | ||
XM_017019906.1 | XP_016875395.1 | 5 | 1680 | ||
XM_017019907.1 | XP_016875396.1 | 6 | 2461 | ||
XM_017019908.1 | XP_016875397.1 | 4 | 1621 | ||
GenBank mRNA | AK312763.1 | - | 4 | 1585 | |
BC065193.1 | - | 4 | 1642 | 5227360 | |
HM013717.1 | - | 3 | 1529 | ||
L31573.1 | - | 3 | 2252 |